Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome Detection (Western Blot) Antibody Pack, Novus Biologicals™
SDP

Catalog No. NB397898 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
9 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397898 9 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397898 Supplier Novus Biologicals Supplier No. NBP311740
Add to Cart
Add to Cart

nan nan Antibody

Exosome Detection (Western Blot) nan antibody specifically detects Exosome Detection (Western Blot) in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Exosome Detection (Western Blot)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Formulation See individual datasheets
Gene Alias EV detection, Exosome Markers, extracellular vesicle detection
Immunogen See individual datasheets
Quantity 9 Vials
Regulatory Status RUO
Target Species Human
Content And Storage Storage is content dependent.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.