Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Exosome Standards (LnCAP cell line)
SDP

Catalog No. NB397931 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397931 6 x 30 μg Vials
NB397928 2 x 30 μg Vials
NB397929 6 x 100 μg Vials
NB397930 2 x 100 μg Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB397931 Supplier Novus Biologicals™ Supplier No. NBP311687180UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

Specifications

Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Quantity 6 x 30 μg Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.