Learn More
Marketlab, Inc. Expandable EZ Lock Dunnage Racks
Description
- Includes two connecting racks
- Interlocking designs allow users to expand their racks to meet their storage needs
- Aerated design allows for better airflow, reducing condensation, bacteria, and dust/debris buildup
- Keeps supplies off the floor to minimize moisture and bacteria growth
- Recycled, high-density polyethylene is freezer safe, chemical resistant, and won't absorb any liquids or chemicals
- Withstands temperatures ranging from -30 to 200°F
- Easy locking system for quick, convenient assembly
- 2,500 lb. load capacity
- Release button for easy disassembly
- 12.75 in. front access; 7 in. side access
Specifications
Specifications
| Size | Standard |
| Product Type | Dunnage Rack |