Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Expanding Our Testing: Using ELISA to Detect COVID-19
SDPSCI_ED

Catalog No. S52002 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S52002 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S52002 Supplier Edvotek™ Supplier No. 263
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this simulated medical test, students will perform the ELISA to detect the presence of COVID-19 antibodies in simulated patient samples.

  • Hands-on simulation of molecular testing for COVID-19
  • Learn about the characteristics of coronaviruses
  • Understand antibodies and immunoassays
  • Perform an ELISA to detect the presence of COVID-19 antibodies in simulated patient samples
  • For 10 Lab Groups
  • Complete experiment in 60 minutes
  • Kit includes 10X ELISA wash buffer, ELISA dilution buffer, antigen (lyophilized), primary & secondary antibodies (lyophilized), ABTS substrate (lyophilized), ABTS reaction buffer, microtiter plates, microcentrifuge tubes, & transfer pipets
  • All you need: Distilled or deionized water, beakers, pipet pump or bulb, lab glassware, disposable lab gloves, & safety goggles. Recommended: automatic micropipettes (0-50 μl, 100-1000 μl) and tips
  • Some components require refrigerator storage
  • Please note, this experiment is a simulation and intended solely for educational purposes. There is no virus or human blood products included with this experiment. The experiment will not detect SARS-CoV-2 antibodies in human blood samples.

Specifications

Grade 8 to Undergraduate
Age 14 to 20
For Use With (Application) Simulated ELISA experiment to detect COVID-19
Includes 10 x ELISA wash buffer, ELISA dilution buffer, antigen (lyophilized), primary & secondary antibodies (lyophilized), ABTS substrate (lyophilized), ABTS reaction buffer, microtiter plates, microcentrifuge tubes, & transfer pipets
Requires Distilled or deionized water, beakers, pipet pump or bulb, lab glassware, disposable lab gloves, & safety goggles. Recommended: automatic micropipettes (0-50 μL, 100-1000 μL) and tips
Content And Storage Some components require refrigerator storage
Class Size 10 Lab Groups
Product Type ELISA Kit

**Please note, this experiment is a simulation and intended solely for educational purposes. There is no virus or human blood products included with this experiment. The experiment will not detect SARS-CoV-2 antibodies in human blood samples.**

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.