Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Expi293™ Expression Medium

Catalog No. A1435104
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1435104 20 L
A1435103 10 L
A1435101 1 L
A1435102 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A1435104 Supplier Gibco™ Supplier No. A1435104
Add to Cart
Add to Cart
Request a Quote

Expi293 Expression Medium designed for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells.

Expi293 Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F cells in the Expi293 Expression System for scalable transient protein expression.

  • Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
  • Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine transfection reagent
  • Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
  • Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10 L in disposable bioreactors
  • Does not contain phenol red

Expi293 Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Expi293F™
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type Expression Medium
With Additives GlutaMAX
Without Additives No Phenol Red
Product Line Expi293
Quantity 20 L
Content And Storage One 20L Universal Bag
Store in the dark at 2-8°C.
Can I purify my protein from Expi293 Expression Medium using ProBond or Ni-NTA purification systems?

Expi293 Expression Medium is not directly compatible with ProBond or Ni-NTA purification systems. We recommend performing a buffer exchange or dialyzing the samples before His-tag purification.

Can I use a different 293 cell line with the Expi293 Expression System?

Other 293 cell lines may be used with the Expi293 Expression System. However, before these cell lines may be used for transfection studies, they must be adapted to serum-free, suspension culture in Expi293 Expression Medium and evaluated for transfection and expression.

Which vector do you recommend using with the Expi293 Expression System?

We recommend using the pcDNA 3.4-TOPO TA vector (Cat. No. A14697). This vector contains the native, full-length CMV promoter and a WPRE (Woodchuck Posttranscriptional Regulatory Element) downstream of the cloning site, both of which contribute to high level gene expression (about 2-3 fold higher expression than with pcDNA3.3 TOPO vector, which in turn provides 2-5 fold higher expression than with a standard pcDNA vector). Of course, the expression level is also protein-dependent.

Is the Expi293 Expression System completely animal-origin free (AOF)?

All the components of the system are animal-origin free except for the Opti-MEM I Reduced Serum Medium that is serum-free but not animal-origin free. Please see the Application Note for using the Expi293 Expression System under animal origin-free conditions:
http://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.5943.file.dat/expi-293-animal-origin-free-co25751.pdf

I am interested in a mammalian expression system that will allow me to obtain very high yields of my protein. What kind of systems do you offer?

For stable high-yield expression, we offer the Freedom CHO-S and Freedom DG44 kits. For transient high-yield expression, we offer the ExpiCHO Expression System, Expi293 Expression System, FreeStyle 293 Expression System, FreeStyle Max 293 Expression System, and FreeStyle Max CHO Expression System. For high-yield expression of functional membrane proteins in Exp293F cells, we offer the Expi293 MembranePro Expression System (Cat. Nos. A25869, A25870) that combines the scalability and ease of use of Expi293 and the technology of MembranePro to allow an increase of more than 20-fold in membrane protein yield compared to the standard, adherent culture MembranePro Functional Protein Expression System. Please see the Application Note (http://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/ProteinExpressionAnalysis/pdfs/PG1391-PJ6257-CO28704-Expi293Adaption-MembraneProTech_AppNoteGlobal-FHR.pdf) for more details.

My Expi293 Expression Medium (Cat. No. A1435102) was accidentally frozen. Can I still use it?

This product should be stored at 2-8 degrees C and should not be frozen. The biggest issue with accidentally freezing this product is its solubility. If this product was accidentally frozen, we recommend placing it in a 2-8 degree environment and allowing it to slowly thaw overnight. If the product is fully thawed and shows no signs of precipitation, then it should still be usable, but we cannot guarantee effectiveness.

Does Expi293 Expression Medium contain antibiotics?

Expi293 Expression Medium does not contain antibiotics. We do not recommend the use of antibiotics, however, 5 mL/L of Antibiotic-Antimycotic (Cat. No. 15240) containing penicillin, streptomycin, and amphotericin B may be used when required, as stated on page 1 in the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/expi293_exp_medium_man.pdf).

Do I need to remove Anti-Clumping Agent and Pluronic F-68 from the medium while growing Expi293 cells? How about during transfection of these cells?

You can have Anti-Clumping Agent and Pluronic F-68 in the medium while growing Expi293 cells. You need to remove Anti-Clumping Agent at least 2 passages prior to transfection of Expi293 cells because it interferes with transfection. You can add it back to the medium after transfection. Pluoronic F-68 can be present during transfection as it does not interfere with transfection.


For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.