Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PureAire Monitoring Systems Explosion Proof Combustible Gas Monitor (LEL)
SDP

Catalog No. 18222016
Change view
Click to view available options
Components Measured:
Ethane, Ethyl Alcohol, Ethylene, Isopropyl Alcohol, Methane, Methyl Alcohol, Xylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
18-222-016 Ethane, Ethyl Alcohol, Ethylene, Isopropyl Alcohol, Methane, Methyl Alcohol, Xylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-222-016 Supplier PureAire Monitoring Systems Supplier No. 95105

PureAire Monitoring Systems Explosion Proof Combustible Gas Monitor (LEL) with a NEMA 4 explosion-proof enclosure

  • Long-life LEL Catalytic Sensor
  • Ideal for facilities that require continuous monitoring of toxic and corrosive gases in hazardous areas
  • In a NEMA 4 is suitable for Class 1, Group B, C, and D, Class 2, Group E, F, and G
  • Used for continuous readings of combustible gases
  • Some detectable gases are hydrogen, natural gas, acetylene, and propane
  • Provides immediate fault identification for a disconnected sensor cell
  • The Air check LEL gas detector is available for hydrogen, natural gas, methane, and more

Specifications

Accuracy ±1% of Reading
Calibration Kit Non-Intrusive
Certifications/Compliance Class 1, Group B, C, D Suitable Enclosure
Depth (English) 7 in.
Depth (Metric) 177.8 mm
Description NEMA 4 Explosion Proof Enclosue
Components Measured Ethane, Ethyl Alcohol, Ethylene, Isopropyl Alcohol, Methane, Methyl Alcohol, Xylene
Dimensions (L x W x H) 4.6 x 5.3 x 6.1 in.
For Use With (Equipment) PureAire Horn and Strobe, Remote Digital Display, 8 Channel Controller
Type Fixed Combustible/Toxic Gas Detector
Display Type 3 1/2 in. Digital LCD
For Use With (Application) Combustible Gases
Humidity Range 5 to 95% RH
Measurement Range 0 to 100%
Outputs 4-20 mA
Temperature Range (English) -25°F to +122°F
Temperature Range (Metric) -32 to +50°C
Voltage 24 V
Weight (English) 4 lb.
Weight (Metric) 1.8 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.