Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PureAire Monitoring Systems Explosion Proof Universal Carbon Monoxide 0-100ppm Gas Monitor
SDP

Catalog No. 17542407
Change view
Click to view available options
Components Measured:
Carbon Monoxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-542-407 Carbon Monoxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-542-407 Supplier PureAire Monitoring Systems Supplier No. 98101
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

PureAire’s EPU Carbon Monoxide Gas Monitor accurately detects low CO levels in PPM. Housed in durable XP-rated casing for hazardous areas, it offers reliable, long-term performance with a set-it-and-forget-it approach—ideal for continuous carbon monoxide monitoring.

  • Detects Carbon Monoxide (CO) 0-100ppm
  • RS-485 Communication Interface
  • 3-5 Year sensor life
  • Easy to use - Set it and forget it technology
  • Operating temperatures: -4°F to +140°F (-20°C to +60°C)
  • Fail-safe - Built-in self-test
  • Back-lit LCD Display
  • 4-20mA output
  • User-adjustable dual alarm relays
  • One fault relay
  • Durable XP Housing; Certified for hazardous locations
  • Haz Loc certified
  • Ratings: Class I and II, Division 1, Groups B, C, D, E, F, G. Class III Type 4X/IP66

Specifications

Components Measured Carbon Monoxide
For Use With (Equipment) Gas Detector
Type Fixed Combustible/Toxic Gas Detector
IP Rating IP66
Voltage 21 to 24 VDC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.