Learn More
Eppendorf 3P Freezer Rack, Upright Slide, 1 Rack/Unit
Supplier:  Eppendorf 3P K0641PRE2
Encompass_Preferred
Drawer or side access. For sample access, racks with drawers stay in the freezer compartment. Only the relevant drawer with the freezer boxes of interest is pulled-out. All other boxes stay in the cold inner area of the ULT freezer. Eppendorf drawers are optimized for smooth movement at -80C for ergonomic handling. When using side access racks, sample access results in removal of most of the storage boxes out of the cold inner area of the ULT freezer. The side access rack has to be pulled out to have access to the samples. Racks for chest freezers can be equipped with locking rods to secure freezer boxes when pulling-out the rack tower. Stainless steel or aluminum. By standard, there are two types of rack materials available.. Stainless steel. Stable/ robust for longevity and higher capacit. For freezer boxes up to 136 mm x 136 mm for high flexibility regarding boxe. Optimized cooling conditions by special design for improved sample safet. Aluminum.
Catalog No. 50-118-0201
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.