Learn More
XpressBio Extended Range HIV-1 p24 ELISA Kit, 96 wells
SDP
Supplier:  XpressBio XBR1000
Encompass_Preferred
ELISA plate wells coated with anti HIV-1 p24 murine monoclonal antibody. The assay is used to monitor HIV-1 in cell culture or to determine the viral titer of lentiviral samples.
Catalog No. NC2050056
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.