Learn More
United Scientific™ Extension Thermometer Clamp
SDPSCI_ED
Catalog No. S03081 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Silver
Catalog No. Color
S03081 Silver
1 options
Prop 65
Catalog No. S03081 Supplier United Scientific™ Supplier No. EXTHCL01
Add to Cart
Add to Cart
Prop 65
Extension Thermometer Clamp ideal for placing thermometer in the center of larger glassware.
This extension clamp holds glass tubing or thermometers securely but gently. Adjustments are made easily with one hand using the wing nut. Clamp opening extends from 6 mm to 16 mm. Jaw width is 15.5 mm. Includes a stainless steel rod that is 15 cm long with a diameter of 10 mm. Overall length is 20 cm. Requires a right-angle clamp to connect to a support rod.
  • All stainless steel construction
  • Holds glass thermometers firmly but gently
  • Easily adjust tension with the included wing nut
Specifications
Type 2-Prong Standard Extension Clamp
Material Nickel-plated
Includes Stainless Steel Rod
Length (English) 7.9 in.
Length (Metric) 20 cm
Color Silver
Jaw Type 2-Prong