Learn More
Brady™ Extra Secure Prolock
Catalog No. 19103913 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Green
Red
Yellow
Includes:
59 in. cable (green)
59 in. cable (red)
59 in. cable (yellow)
Without Cable
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
19-103-913 Yellow 59 in. cable (yellow)
19-103-915 Red Without Cable
19-103-916 Yellow Without Cable
19-103-917 Green Without Cable
19-103-914 Green 59 in. cable (green)
19-103-912 Red 59 in. cable (red)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-103-913 Supplier Brady™ Supplier No. 122242
Effectively lock out a variety of isolation points. The Brady™ Extra Secure Prolock will quickly become a staple lockout device in any facility. It tackles anything from T valve handles to electrical disconnects.
  • Designed to effectively lock out a variety of isolation points, including gate valves, T valve handles, electrical disconneccts, and more
  • Economical and commact devices are highly visible and will quickly become a staple lockout device in any facility
Specifications
Product Type Pull-tight Cable Lockout Device
Color Yellow
Includes 59 in. cable (yellow)
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.