Learn More
Abcam Extracellular Vesicle Membrane Staining Kit, 560/585 nm, Fluorescent, 500 Tests
Description
- Assay type: Cell-based
- Applications: Flow Cyt
- Detection method: Fluorescent
- Reactive species: Mammals
- Sample types: Adherent cells, Suspension cells
- Results type: Quantitative
- Efficiency: Labeling in < 4 hrs with 30 secs hands-on time; 100% antibody recovery
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Extracellular Vesicles |
| Reactivity | Mammals |
| For Use With (Equipment) | Flow Cytometer |
| No. of Reactions | 500 Tests |
| Content And Storage | Store at -20°C. |
| Format | Kit |
| Detection Method | Fluorescence |
| Cross Reactivity | Mammals |
| Sample Type | Adherent cells, Suspension cells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.