Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Extraction Buffer, 15 mL

Catalog No. 502663122
Encompass_Preferred
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-266-3122 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-266-3122 Supplier Abcam Supplier No. AB26049015ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-quality extraction buffer for use in sample preparation and detection kits.

  • Product type: Extraction buffer
  • Range: Sample prep and detection kits

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Wash Solution
Format Bottle
Kit Contents 15 mL Extraction Buffer
Quantity 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.