Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EYA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113674 Supplier Thermo Scientific Supplier No. PA5113674
Only null left
Add to Cart
Add to Cart

EYA1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen EYA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EYA1
Gene Accession No. Q99502
Gene Alias BOP; bor; BOS1; dog eared; EYA transcriptional coactivator and phosphatase 1; eya1; eya1 protein; Eyes absent; eyes absent 1; eyes absent 1 homolog; eyes absent 1 homolog (Drosophila); eyes absent homolog 1; MGC141875; OFC1; OTTHUMP00000195053; wu:fc13c10; zgc:100770
Gene Symbols EYA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human EYA1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2138
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.