Learn More
Eye Wonder Book Series

Description
- Reference series specially developed for young students ready to read about the natural world
- Written in text that is clear, understandable, informative
- Photography depicts subjects in their natural settings
- Hardcover; 48 pages; Grades K-3
Bugs
Explores the world of insects from dung beetles to dragonflies and more. 2002.
Human Body
Brief explanation of the human body; also includes some facts about sleep. 2003.
Dinosaur
Introduces students to many types of dinosaurs such as the Tyrannosaurus Rex, the Velociraptor, and the Hadrosaur. 2001.
Earth
Presents general information about the earth with high-quality images. 2002.
Human Body
Brief explanation of the human body; also includes some facts about sleep. 2003.
Ocean
Describes coral reefs, kelp forests, and other underwater worlds, animals such as whales, dolphins, sharks, and more. 2001.
Space
Guide to outer space and astronomy that includes information about exploration, solar system, planets, stars, and galaxies. 2001.
Rocks and Minerals
Teaches geology topics to young students such as the characteristics of rocks and minerals. 2003.
Volcano
Answers questions like How are volcanoes formed? and How hot is molten lava? Also presents information about famous volcanic eruptions. 2003.
Specifications
Specifications
| Grade | K to 7 |
| Format | Book |
| Product Type | Ocean |
| For Use With (Application) | Describe coral reefs, kelp forests and other underwater worlds, animals such as whales, dolphins, sharks and more |
| Subject | Geography |
| Author | Samantha Gray |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.