Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EYFP-Tag Monoclonal Antibody (1H9), Bioss
SDP

Catalog No. 502281854
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-228-1854 Supplier Bioss Supplier No. BSM33182M
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EYFP-Tag Monoclonal antibody specifically detects EYFP-Tag in Tag samples. It is validated for Western Blot

Specifications

Antigen EYFP-Tag
Applications Western Blot
Classification Monoclonal
Clone 1H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene Alias GFP; Yellow fluorescent; YFP
Host Species Mouse
Immunogen Recombinant EYFP protein
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.