Learn More
Zymo Research Corporation EZ-96 DNA Methylation-Gold™ Kit (Shallow-Well) (2 x 96 Rxns)
SDP
Supplier:  Zymo Research Corporation D5007
Encompass_Preferred
The EZ-96 DNA Methylation-Gold Kit allows high-throughput (96-well spin-plate) conversion of unmethylated cytosines into uracil in less than 3 hours. Our optimized bisulfite chemistry combines heat denaturation and bisulfite conversion of input DNA to reduce incubation time and ensure conversion rates >99%. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-203-7108
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.