Learn More
Zymo Research Corporation EZ-96 DNA Methylation-Lightning MagPrep (CE-IVD), 4 x 96 rxns.
SDP
Supplier:  Zymo Research Corporation D5046E
Encompass_Preferred
The EZ-96 DNA Methylation-Lightning MagPrep Kit is the fastest magnetic bead-based DNA bisulfite conversion kit for high-throughput and automated methylation analysis. No preparation is necessary with the ready-to-use conversion reagent - simply add this unique reagent to a DNA sample and incubate the reaction for about one hour. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. This magnetic bead-based bisulfite conversion kit produces the highest yields of bisulfite-converted DNA that is ideal for PCR, arrays, library preps, and Next-Generation sequencing.
Catalog No. NC2134222
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.