Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-Accu Shot™ - Burkholderia Cenocepacia
SDP

Catalog No. 23004001 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-004-001 5 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-004-001 Supplier Microbiologics™ Supplier No. 01269A
Add to Cart
Add to Cart

Microbiologics™ EZ-Accu Shot - Burkholderia Cenocepacia is a fast and reliable solution for Growth Promotion or Media Challenge Testing

  • EZ-Accu Shot provides a fast, convenient and reliable solution for Growth Promotion Testing, Media Challenge Testing and more
  • Each instant-dissolve microorganism pellet is designed to deliver 10-100 CFU per inoculum
  • Provides 8 hours of stability after rehydration, meeting Pharmacopeia guidelines with ease
  • Quantitative microorganism preparation delivers <100 CFU per 0.1 mL of hydrated suspension
  • Kit includes: 5 pellets of a single enumerated strain, and 5 vials of hydrating fluid (1.2 mL per vial)
  • No dilutions required

Specifications

ATCC® ATCC BAA-245
Biosafety Level 2
Content And Storage 2°C to 8°C
Format EZ-Accu Shot
Manufacturing Origin United States
Product Type Quality Control
For Use With (Application) Growth Promotion Testing
Packaging Type Plastic Box
Quantity 5 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.