Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-Accu Shot™ Starved Cells
SDP

Catalog No. 23004002 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Methylobacterium extorquens
Pseudomonas protegens
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-004-002 Methylobacterium extorquens
23-004-003 Pseudomonas protegens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-004-002 Supplier Microbiologics™ Supplier No. 01110SC
Add to Cart
Add to Cart

Microbiologics™ EZ-Accu Shot™ Starved Cells simplify and accelerate test procedures for growth promotion of R2A agar

  • Laboratories following Japanese Pharmacopeia (JP) guidelines know the challenges of performing Media Growth Promotion Testing on R2A agar
  • The JP states that R2A media should be challenged with strains that have been starved for 3 days prior to conducting the test
  • To simplify and accelerate test procedures for growth promotion of R2A agar, we have developed EZ-Accu Shot™ Starved Cells
  • These test-ready control strains have been starved prior to lyophilization and deliver the required concentration of 50-200 CFU per 1 mL inoculum on R2A media
  • Designed for laboratories performing the Growth Promotion Test of R2A media according to the Japanese Pharmacopeia (JP)
  • No dilutions required
  • Kit includes: 5 vials of a single, quantitative microorganism (1 lyophilized pellet per vial)

Specifications

ATCC® ATCC BAA-2500
Biosafety Level 1
Content And Storage 2°C to 8°C
Format EZ-Accu Shot Starved Cells
Manufacturing Origin United States
Product Type EZ-Accu Shot
For Use With (Application) Growth Promotion Testing, Pharmaceutical Microbial Enumeration Testing, Sterility Testing
Packaging Type Plastic Box
Quantity 5 Pellets
Species Methylobacterium extorquens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.