Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MicroBioLogics™ EZ-Accu Shot™ Burkholderia cepacia Derived from ATCC™ 25416™†
SDP

Catalog No. 16230463 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Burkholderia cepacia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
16-230-463 Burkholderia cepacia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-230-463 Supplier MicroBioLogics™ Supplier No. 0488A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EZ-Accu Shot for fast, reliable Growth Promotion Testing

  • Quantitative microorganism preparation that delivers less than 100 CFU per 0.1mL of hydrated suspension
  • Ready-to-use, no pre-incubation
  • Instant dissolve pellet, no dilutions required
  • Up to 8 hours stability for ultimate flexibility
  • Strains are four passages or fewer from reference culture
  • Traceability to reference culture ensures authenticity
  • Online Certificate of Analysis
  • Biosafety Level: 2
  • Kit contains: 5 pellets of a single enumerated strain and 5 vials of hydrating fluid (1.2mL per vial).

Specifications

ATCC® 25416
Biosafety Level 2
Format EZ-Accu Shot™
For Use With (Application) Growth Promotion Testing
Includes 5 pellets of a single enumerated strain and 5 vials of hydrating fluid (1.2 mL per vial)
Species Burkholderia cepacia

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics™ is licensed to use these trademarks and to sell products derived from ATCC™ cultures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.