Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EZ-ChIP™

Catalog No. 17371 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
22 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-371 22 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-371 Supplier MilliporeSigma™ Supplier No. 17371
Add to Cart
Add to Cart

Includes

  • DNA Purification Spin Columns and Collection Tubes
  • ChIP Blocked Protein G Agarose
  • Anti-RNA Polymerase II
  • Control PCR Primers
  • Normal Mouse IgG
  • Bind, Wash and Elution Reagents
  • Protease Inhibitor Cocktail II
  • RNase A
  • Proteinase K
  • All required buffers
  • ChIP Dilution Buffer
  • Low Salt Immune Complex Wash Buffer
  • High Salt Immune Complex Wash Buffer
  • LiCl Immune Complex Wash Buffer
  • TE Buffer
  • 0.5M EDTA
  • 5M NaCl
  • SDS Lysis Buffer
  • 1M Tris-HCl, pH 6.5
  • 10X PBS
  • 10X Glycine
  • 1M NaHCO3
  • Control Primers
  • 20% SDS
  • Spin Filters
  • Collection Tubes
  • Bind Reagent A
  • Wash Reagent B
  • Elution Reagent C

Any chromatin-associated or DNA binding protein can be analyzed with this technique, provided a good antibody to the protein exists

  • Easier: Spin columns make DNA purification easier and more reliable - no more messy phenol-chloroform extractions
  • Quicker: All reagents to process your samples are included - you don't have to spend valuable time making them
  • Greater Reproducibility: Positive and negative control antibodies and PCR primers are included to help validate your results and to troubleshoot your experiments
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein G agarose beads
Quantity 22 Assays
Stability Stable for 1 year from date of shipment when stored as directed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.