Learn More
Air-Tite EZ-Dose Syringe
GREENER_CHOICE
Catalog No. 14818151 Shop All Air-Tite Products
Change view
Click to view available options
Volume (Metric):
1 mL
10 mL
20 mL
3 mL
5 mL
50 to 60 mL
Connection Type:
Centric Luer Lock
Centric Luer Slip
Eccentric Luer Slip
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connection Type
14-818-151 1 mL Centric Luer Lock
06-480-IMI 5 mL Centric Luer Slip
14-818-152 1 mL Centric Luer Slip
14-818-147 3 mL Centric Luer Lock
14-818-148 3 mL Centric Luer Slip
14-818-155 5 mL Centric Luer Lock
14-818-156 5 mL Centric Luer Slip
14-818-146 10 mL Centric Luer Lock
14-818-145 10 mL Centric Luer Slip
14-818-149 20 mL Centric Luer Lock
14-818-150 20 mL Eccentric Luer Slip
14-818-153 50 to 60 mL Centric Luer Lock
14-818-154 50 to 60 mL Eccentric Luer Slip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 14-818-151 Supplier Air-Tite Supplier No. MEZ1LL
Add to Cart
Add to Cart
Built for precision and reliability, EZ-Dose Syringes offer exceptional quality and durability.
Designed exclusively for laboratory and veterinary use, EZ-Dose Syringes come in eco-friendly, fully recyclable packaging, delivering performance you can trust, with sustainability in mind.
Specifications
Autoclavable Not Autoclavable
Material Polypropylene (Barrel), Silicone Oil (Lubricant), Polypropylene (Plunger), Polyisoprene Rubber (Tip), Polypropylene (Body)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Disposable Syringe
Disposable Single-use
Volume (Metric) 1 mL
For Use With (Equipment) Hypodermic Needle, Safety Needle, Blunt Fill Needle
Connection Type Centric Luer Lock
Graduated Yes
Sterility Sterile