Description
Designed exclusively for laboratory and veterinary use, EZ-Dose Syringes come in eco-friendly, fully recyclable packaging, delivering performance you can trust, with sustainability in mind.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Polypropylene (Barrel), Silicone Oil (Lubricant), Polypropylene (Plunger), Polyisoprene Rubber (Tip), Polypropylene (Body) |
| Needle Gauge | No Needle Included |
| Latex Free | Yes |
| Product Type | Disposable Syringe |
| Disposable | Single-use |
| Volume (Metric) | 1 mL |
| For Use With (Equipment) | Hypodermic Needle, Safety Needle, Blunt Fill Needle |
| Connection Type | Centric Luer Slip |
| Graduated | Yes |
| Show More |