Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Biotin-PEG12-DBCO

Catalog No. FERC20042 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERC20042 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERC20042 Supplier Thermo Scientific™ Supplier No. C20042
Add to Cart
Edge
Add to Cart

EZ-Link Biotin-PEG12-DBCO is a biotinylation reagent that contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry attachment to an azide-labeled molecule, enabling the attachment of biotin using a bioorthogonal approach.

EZ-Link Biotin-PEG12-DBCO is a biotinylation reagent that contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry attachment to an azide-labeled molecule, enabling the attachment of biotin using a bioorthogonal approach. The DBCO group reacts preferentially with an azide through a copper-free strain-promoted azide-alkyne cycloaddition (SPAAC) reaction to form a stable triazole linkage with an azide-labeled coupling partner. EZ-Link Biotin-PEG12-DBCO contains a 12-unit polyethylene glycol spacer arm for increased water solubility and reach.

EZ-Link Biotin-PEG12-DBCO features include:
• Bioorthogonal—DBCO and azides react selectively to form triazole linkages
• Enhanced solubility—pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
• Irreversible—forms irreversible and stable triazole linkage

Classic click reactions involve a copper-catalyzed azide-alkyne cyclo­addition to label or conjugate molecules. A drawback of this approach is that copper ions can impair protein function, harm cells, and reduce the fluorescence of fluorophores. DBCO is distinct in that it doesn’t require copper to catalyze the reaction with an azide for triazole formation. The strain in the eight-membered ring allows the reaction with azide-modified molecules to occur in the absence of catalysts, preventing copper-induced damage to cells or proteins.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Azide
Cleavable No
Spacer Arm Length 46.3 Å
Content And Storage Store desiccated at –20°C
Shipping Condition Wet Ice
Product Line EZ-Link
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety DBCO, Biotin
Spacer Long (>30 Å)
Quantity 10 mg
Solubility DMSO or DMF
Format Standard
Product Type Biotin-PEG12-DBCO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.