Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin
Description
Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
- Protein labeling - biotinylate antibodies or other proteins for detection or purification using streptavidin probes or resins
- Amine-reactive - reacts with primary amines (-NH2), such as the side-chain of lysines (K) or the amino-termini of polypeptides
- Pegylated - spacer arm contains a hydrophilic, 4-unit, polyethylene glycol (PEG) group
- Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
- Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
- Long reach - spacer arm (total length added to target) is 29 angstroms; this reduces steric hindrance when binding to avidin molecules
Specifications
Specifications
| Description | EZ-Link NHS-PEG4-Biotin |
| Product Type | NHS-PEG4-Biotin |
| Quantity | 1 g |
| Content And Storage | Store at -20°C. |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Impermeant |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
| Label or Dye | Biotin |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.