Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ TFP Ester-PEG12-DBCO

Catalog No. FERC20040 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERC20040 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERC20040 Supplier Thermo Scientific™ Supplier No. C20040
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A heterobifunctional crosslinker that contains an amine reactive tetrafluorophenyl (TFP) and a dibenzylcyclooctyne (DBCO) with a water solubilizing long PEG12 spacer in between. Use to label any primary amine containing molecule such as a protein to biomolecule with a distantly attached DBCO group.

  • A heterobifunctional crosslinker that contains an amine reactive tetrafluorophenyl (TFP) and a dibenzylcyclooctyne (DBCO) with a water solubilizing long PEG12 spacer in between the two groups
  • This crosslinker can be used to label any primary amine containing molecule such as a protein to biomolecule with a distantly attached DBCO group
  • Once a protein or (bio)molecule is DBCO labeled, it can undergo a copper-free azide-alkyne cycloaddition reaction to form a stable triazole linkage with an azide labeled coupling partner
  • Can be used to create diverse conjugates with the biorthogonal coupling between an alkyne and azide (supplied separately)
  • The coupling or crosslinking procedure is typically done in two steps:
    • First, a protein or amine containing molecule is labeled with a DBCO group and excess crosslinker is removed
    • Second, an azide containing moiety (prepared independently) is introduced into the same system as the DBCO moiety
    • Labeling occurs under biological conditions without the introduction of any additional reagents such as a copper catalyst
  • Reagent readily dissolves in water-miscible solvents for dilution into aqueous reaction mixtures
  • TFP esters are more hydrolytically stable than NHS esters
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Azide
Cleavable No
Description TFP Ester-PEG12-DBCO
Molecular Weight (g/mol) 1067.12
PEGylated Yes
Spacer Arm Length 46.3 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability Not determined
Shipping Condition Wet Ice
Product Line EZ-Link
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety TFP Ester, DBCO
Spacer Long (>30 Å)
Form Liquid
Quantity 25 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.