Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EZ-Magna ChIP™ A/G Chromatin Immunoprecipitation Kit

Catalog No. 1710086 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
22 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-100-86 22 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-86 Supplier MilliporeSigma™ Supplier No. 1710086
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Magnetic Protein A/G Beads
  • ChIP Dilution Buffer
  • Low Salt Wash Buffer
  • High Salt Wash Buffer
  • LiCl Wash Buffer
  • TE Buffer
  • Cell Lysis Buffer
  • Nuclear Lysis Buffer
  • ChIP Elution Buffer (w/o Proteinase K)
  • Proteinase K
  • RNase A
  • 10X Glycine
  • 10X PBS
  • Protease Inhibitor Cocktail II
  • Anti-RNA polymerase II, clone CTD4H8
  • Normal Mouse IgG
  • Control Primers
  • Spin Filters
  • Collection Tubes
  • Bind Reagent A
  • Wash Reagent B
  • Elution Reagent C

Requires

Magna Grip Rack 8 well

Analyze changes in histone modifications that correlate with processes like transcription, mitosis or DNA repair

  • Magnetic bead-based rapid protocol allows performance of ChIP in as little as 1 day
  • Blend of protein A+G bead blend allows ChIP using a broader range of antibodies than A or G alone
  • Negative and positive control antibodies and control primer set to simplify validation of experimental procedure
  • Includes spin columns to make DNA purification easier and more reliable
  • Complete kit with all required reagents for reliable and reproducible results
  • Compatible with ChIPAb+ validated antibody and primer sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) To perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads
Quantity 22 Assays
Stability Kit components are stable for 1 year from date of shipment when stored as directed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.