Learn More
Marketlab, Inc. EZ-Nest Sample Cups
Description
- Lower shoulder height prevents jamming of the instrument sample arm against the cup
- Top flange prevents cup from being immersed too far
- High clarity polystyrene
Specifications
Specifications
| Capacity (Metric) | 1 mL |
| Product Type | Nesting Sample Cup |
| Capacity (English) | 0.0338 oz. |
| Material | Polystyrene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.