Learn More
PIP™ EZ-Twist™ Soft Polyurethane Foam Ear Plugs
Catalog No. 19046466 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Corded:
Corded
Uncorded
Pairs per Package:
100 pairs/dispenser box
200 pairs/dispenser box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded Pairs per Package
19-046-466 Uncorded 200 pairs/dispenser box
19-046-465 Corded 100 pairs/dispenser box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-046-466 Supplier PIP™ Supplier No. 267HPF610
Add to Cart
Add to Cart
EZ-Twist™ push-in ear plugs conform to most ear canals.
  • Just insert and twist
  • Shape conforms to most ear canals
  • NRR 30 protection
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Oil and gas, Manufacturing, Workshops, Building, Construction, Energy
High Visibility Yes
Packaging Type Poly bag
Pairs per Package 200 pairs/dispenser box
Plug Material Polyurethane foam
Plug Color Orange
Noise Reduction Rating (NRR) 30 dB
Corded Uncorded
Plug Shape Cylinder
Product Type Disposable Ear Plugs
Disposable Single-use
Metal Detectable No
Dielectric Yes
EN 352.2002 EN 352-2