Learn More
Restek™ EZ Twist Top Injection Port Kits
SDP
Change inlet liners faster and easier, for Agilent 5890/6850/6890/7820 GCs
Supplier:  Restek™ 22735
 View more versions of this product
Catalog No. 06-611-242
Description
Description
  • Gas lines are attached to shell weldment bottom to help eliminate broken gas lines
  • Gas lines do not interfere with routine maintenance
  • Weldment Removal Tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces
  • Weldment stays secure in the tool for reattachment
Specifications
Specifications
Injection Port Kit
EZ Twist Top
Agilent 6850/6890/7820 (Large Canister Type Filter)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.