Learn More
Restek™ EZ Twist Top Injection Port Kits
SDP
Catalog No. 06611243 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Injection Port Kit
Siltek™ Injection Port Kit
For Use With (Equipment):
Agilent 5890
Agilent 6850/6890/7820
Agilent 6850/6890/7820 (Large Canister Type Filter)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
06-611-243 Siltek™ Injection Port Kit Agilent 6850/6890/7820 (Large Canister Type Filter)
06-611-235 Siltek™ Injection Port Kit Agilent 5890
06-611-230 Injection Port Kit Agilent 6850/6890/7820
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-611-243 Supplier Restek™ Supplier No. 22736
Change inlet liners faster and easier, for Agilent 5890/6850/6890/7820 GCs
  • Gas lines are attached to shell weldment bottom to help eliminate broken gas lines
  • Gas lines do not interfere with routine maintenance
  • Weldment Removal Tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces
  • Weldment stays secure in the tool for reattachment
Specifications
Product Type Siltek™ Injection Port Kit
Description EZ Twist Top
For Use With (Equipment) Agilent 6850/6890/7820 (Large Canister Type Filter)