Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EZ-Zyme™ Chromatin Prep Kit

Catalog No. 17375 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-375 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-375 Supplier MilliporeSigma™ Supplier No. 17375
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Store at 4°C:

  • 0.5M EDTA
  • 5M NaCl
  • 1M Tris-HCl, pH 6.5
  • 10X Glycine
  • 10X PBS
  • EZ-Zyme™ Lysis Buffer
  • EZ-Zyme™ Digestion Buffer
  • EZ-Zyme™ Stop Buffer

Store at -20°C:

  • Protease Inhibitor Cocktail II.
  • RNase A
  • Proteinase K
  • EZ-Zyme™ Enzymatic Cocktail

Requires

  • Cells, stimulated or treated as needed for the experimental system
  • 37% Formaldehyde
  • Liquid nitrogen (N2 ) or dry ice/alcohol bath
  • Note: care should be exercised in handling liquid nitrogen as liquid can damage skin or tissue on contact. Use appropriate personal protective equipment when working with liquid nitrogen or the dry ice/alcohol bath

ChIP analysis at nucleosome resolution by performing complete or partial digestions with a proprietary enzymatic cocktail to obtain chromatin fragments of on average one to a few nucleosomes in length

  • ChIP analysis at nucleosome resolution by performing complete or partial digestions with a proprietary enzymatic cocktail to obtain chromatin fragments of on average one to a few nucleosomes in length.
  • Compared to sonication, this procedure not only allows mapping the protein-DNA association at a higher resolution, but also renders the subsequent immunoprecipitation more efficient due to its mild digestion condition. 
  • In addition, enzymatic shearing is compatible with non-crosslinked ChIP (native ChIP), which allows analysis of histone modifications not only on cultured cells, but also on freshly dissected and frozen tissues 
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) Enzymatic shearing of chromatin from mammalian cells at higher resolution than sonication for use in chromatin immunoprecipitation (ChIP) assays
Quantity 1 Kit
Stability Stable for 6 months from date of shipment when stored as directed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.