Learn More
Foxx Life Sciences EZBio Rectangular Carboy with 4" TC Sanitary Neck, No Spigot, Polypropylene (PP), 2.5L Carboy with Handle, Powder Carboy, Autoclavable, Non-Sterile, 1/CS
Supplier:  Foxx Life Sciences 5155C82FLS
Encompass_Preferred
EZBio Carboy with Sanitary Neck is a fluid and powder handling container designed for bioprocessing, laboratory, and pharmaceutical applications. Designed for closed-system fluid, it features 4” and 6” tri-clamp sanitary necks, ensuring contamination-free connections in cleanroom environments. Available in round and rectangular formats, the round sanitary carboy supports efficient mixing, while the rectangular design provides space-saving storage. Constructed from high-purity plastic this tri-clamp carboy delivers reliable performance for bioprocess production, buffer and media preparation, sampling, and waste collection—making it a trusted solution across upstream and downstream workflows.
Features
4” and 6” tri-clamp sanitary necks for closed-loop operation
Materials comply with USP Class VI standards
BPA-free, gamma-sterilizable
Wide-mouth ergonomic design for easy filling and cleaning
Ideal for buffer/media prep, powder transfer, and waste collection
Catalog No. 50-003-04234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.