Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188356
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2612 10 μL
50-199-3521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2612 Supplier Active Motif Supplier No. 61584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 ).

Specifications

Antigen EZH1
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene EZH1
Gene Accession No. P70351
Gene Alias BOS_19112; enhancer of zeste 1 polycomb repressive complex 2 subunit; Enhancer of zeste homolog 1; enhancer of zeste homolog 1 (Drosophila); Enx2; ENX-2; Ezh1; Histone-lysine N-methyltransferase EZH1; hypothetical protein LOC664754; KIAA0388; KMT6B; si:ch211-250i3.1; zgc:136829
Gene Symbols EZH1
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the N-terminal region of human EZH1.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14055
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.