Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eZKine™ CD8 Activation 1 Whole Blood Intracellular Cytokine Kit

Catalog No. 8822685441
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-226-85441 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-226-85441 Supplier Invitrogen™ Supplier No. 8822685441

This eZKine CD8 Activation 1 Whole Blood Intracellular Cytokine Kit is designed to rapidly identify cytokine producing T lymphocytes of the CD8+ T cell lineage after stimulation of whole peripheral blood samples.

This eZKine CD8 Activation 1 Whole Blood Intracellular Cytokine Kit is designed to rapidly identify cytokine producing T lymphocytes of the CD8+ T cell lineage after stimulation of whole peripheral blood samples. Stimulation of whole blood in the presence of Protein Transport Inhibitor Cocktail (cat. 00-4980, not included) is followed by red blood cell lysis and fixation in a single step and subsequent permeabilization. Samples are then ready to stain with the CD8 Activation 1 cocktail and concentration-matched Isotype Control cocktail.

CD8+ T cells, or cytotoxic T lymphocytes, are immune cells that are responsible for the detection and clearance of virally or bacterically infected host cells. Upon recognition of antigen displayed on the surface of infected cells, activated CD8+ T cells upregulate markers of early activation such as CD69 and FAS ligand, followed by the release of granules containing granzymes and perforin as well as secretion of cytokines such as IFN gamma and TNF alpha. Granzymes and perforin act in concert to directly lyse infected target cells, while cytokines have paracrine effects including the recruitment and activation of macrophages, lymphocytes, and PMNs. IFN gamma is important for the upregulation of MHC class I and II on nearby cells as well as promoting Th1 cell differentiation.

  • Reactivity/Species: Human
  • Reported Application: Intracellular Staining followed by Flow Cytometric Analysis
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
No. of Tests 25 tests
Quantity 25 tests
For Use With (Application) Intracellular Staining, Flow Cytometry
Product Line eZKine
Product Type Intracellular Cytokine Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.