Learn More
Omega Biotek Inc EZNA DNA KIT 200 PREP
Supplier: Omega Biotek Inc D339602
            The E.Z.N.A.� Tissue DNA Kit offers a simple, rapid, and cost-effective method for the isolation of DNA from a wide variety of sample sources include fresh or frozen animal cells and tissues. After cell lysis, the DNA purification process can be completed in fewer than 30 minutes. Up to 30 mg of tissue at a time can be easily processed using the simple E.Z.N.A.� Tissue DNA protocol. With the spin-column based kit, multiple samples can be processed in parallel. There is no need for phenol/chloroform extractions or time-consuming steps such as precipitation with isopropanol or ethanol. DNA purified using the E.Z.N.A.� Tissue DNA Kit is ready for most downstream applications such as PCR, Southern blots, and restriction enzyme digestion.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.