Learn More
Sigma Aldrich Fine Chemicals Biosciences EZVIEW RED ANTI-FLAG M2,1ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F24261ML
Encompass
EZview Red anti-flag M2 affinity GelM2, monoclonalEZ view Red Anti-FLAG M2 Affinity Gel is a resin that consists of Anti-FLAG M2 antibody, covalently bonded to 4% Agarose beads. The affinity gel is used to bind FLAG fusion proteins to samples, such as cell lysates and tissue, for purification of FLAG-tagged proteins in preparation of immunoprecipitation assays. Red dye enhances visability for more efficient results. Agarose beads bind at N-terminal, Met-N-terminal and C-terminal FLAG fusion proteins, 3xFLAG-tagged fusion proteins.
Catalog No. NC1845148
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.