Learn More
Description
Specifications
Specifications
| Packaging | Ampule |
| Shipping Condition | Dry Ice |
| Physical Form | Neat |
| Synonym | AOCS Reference Mixtures; AOCS No. 4 (RM-4); Oil Reference Standard, AOCS No. 4; Oil Reference Standard, AOCS |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Grade | Certified Reference Material |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.