Learn More
Invitrogen™ F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Functional Grade, eBioscience™
F(ab')2-Goat Polyclonal Secondary Antibody
$391.00
Specifications
Antigen | Mouse IgG (H+L) |
---|---|
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Functional Assay, Flow Cytometry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-9233
![]() |
Invitrogen™
16509885 |
500 μg |
Each of 1 for $391.00
|
|
|||||
Description
Description: This polyclonal F(ab')2 fragment recognizes mouse IgG. Reactivity to human, bovine, and horse has been removed. This polyclonal antibody can be used to crosslink and activate B lymphocytes. Applications Reported: This polyclonal F(ab')2 Goat Anti-Mouse IgG has been reported for use in functional assays (such as B cell activation). Concentrations should be determined empirically for optimal performance in the assay of interest. Applications Tested: F(ab')2 Anti-Mouse IgG Functional Grade Purified has been tested by in vitro stimulation of mouse splenocytes. Endotoxin: Less than 0.1 ng/μg antibody as determined by the LAL assay. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgG (H+L) | |
4°C | |
Polyclonal | |
Liquid | |
Ig | |
Mouse | |
Secondary | |
Affinity chromatography |
1 mg/mL | |
Functional Assay, Flow Cytometry | |
Functional Grade | |
Goat | |
RUO | |
PBS with no preservative; pH 7.2 | |
Secondary Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.