Learn More
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Qdot™ 655
F(ab')2-Goat Polyclonal Secondary Antibody
$332.00 - $394.65
Specifications
Antigen | Rabbit IgG (H+L) |
---|---|
Content And Storage | 4°C, store in dark |
Applications | Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Qdot 655 |
Description
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rabbit IgG (H+L) | |
Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
Qdot 655 | |
Goat | |
RUO | |
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
Secondary | |
purified |
4°C, store in dark | |
Polyclonal | |
Liquid | |
IgG | |
Rabbit | |
Gamma Immunoglobins Heavy and Light chains. | |
Secondary Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.