Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ F-actin Monoclonal Antibody (NH3)
GREENER_CHOICE

Catalog No. MA180729 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA180729 500 μg
MA180163 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA180729 Supplier Invitrogen™ Supplier No. MA180729
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Mouse anti Human F-Actin antibody, clone NH3 recognizes human Filamentous actin (F-actin).

This gene encodes one of six different actin proteins. Actins are highly conserved proteins that are involved in cell motility, structure, and integrity. This actin is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen F-actin
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone NH3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ACTB
Gene Accession No. P29751, P60709, P60710, P60711
Gene Alias ACT; ACT-5; ACTB; ACTG; ACTG1; actin beta; actin gamma 1; actin, beta; actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; Actx; Atcb; beta cytoskeletal actin; beta-actin; BRWS1; BRWS2; cytoplasmic beta-actin; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; E430023M04Rik; epididymis luminal protein 176; gamma non-muscle actin; Gamma-actin; HEL-176; PS1TP5-binding protein 1; PS1TP5BP1
Gene Symbols ACTB
Host Species Mouse
Immunogen Human monocytes and U937 cell line.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009272, 11461, 60, 81822
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.