Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-Actin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983918
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3918 Supplier Bioss Supplier No. BS1571R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes one of six different actin proteins. Actins are highly conserved proteins that are involved in cell motility, structure, and integrity. This actin is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins.

Specifications

Antigen F-Actin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ACTB
Gene Accession No. P60709
Gene Alias ACT-5; ACTB; actin beta; actin, beta; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actx; Atcb; beta cytoskeletal actin; Beta-actin; BRWS1; cytoplasmic beta-actin; E430023M04Rik; gamma non-muscle actin; PS1TP5-binding protein 1; PS1TP5BP1
Gene Symbols ACTB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human F-actin capping protein alpha subunit.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.