Learn More
Abcam F-actin Staining Kit, Green Fluorescence, Cytopainter, 500 Test
Description
- Assay type: Cell-based
- Sample types: Tissue, Suspension cells, Adherent cells
- Results type: Qualitative
- Compatible with: Fluorescence microscope
- Cited in: 18 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | F-actin |
| Detection Location | In-Cell Detection |
| For Use With (Equipment) | Fluorescence Microscope |
| Format | Kit |
| Detection Method | Fluorescence |
| No. of Reactions | 500 Tests |
| Sample Type | Tissue, Suspension Cells, Adherent Cells |
| Quantity | 500 Tests |
| For Use With (Application) | F-actin staining |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.