Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam F-actin Staining Kit, Red Fluorescence, 500 Tests

Catalog No. 502584946
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4946 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4946 Supplier Abcam Supplier No. AB112127500TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized cell-based assay kit for fluorescent staining of F-actin in tissue and cell samples.

  • Stains beta Actin in cell or tissue extracts and cell samples
  • Suitable for use with fluorescence microscope
  • Assay type: Cell-based
  • Sample types: Tissue, suspension cells, adherent cells
  • Cited in 22 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule F-actin
For Use With (Equipment) Fluorescence Microscope
No. of Reactions 500 Tests
Detection Location In-Cell Detection
Format Kit
Detection Method Fluorescence
Sample Type Tissue, Suspension Cells, Adherent Cells
Quantity 500 Tests
For Use With (Application) Immunocytochemistry, Immunofluorescence
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.