Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ F. keratolyticus Endo-beta-Galactosidase Protein

Catalog No. 8620GH050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-20G-H050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 86-20G-H050 Supplier R&D Systems™ Supplier No. 8620GH050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave keratan sulfate. 50% cleavage of keratan sulfate (0.5 μg) is achieved using <5 ng rF.k. Endo-beta-galactosidase, as measured under the described conditions.

Specifications

For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Molecular Weight (g/mol) M.W. (Observed): 66 kDa, reducing conditions; M.W. (theoretical): 59 kDa
Quantity 50 μg
Source E. coli-derived f. keratoyticus Endo-beta-Galactosidase protein msyB (Thr2-Arg124) (N-terminus) Accession # P25738 GSENLYFQGHMHHHHHH F. keratolyticus Endo-beta-Galactosidase (Asp25-Leu422) (N-terminus) Accession # Q9ZG90 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Common Name Endo-beta-Galactosidase
Biological Activity Measured by its ability to cleave keratan sulfate. 50% cleavage of keratan sulfate (0.5μg) is achieved using <5 ng rF.k. Endo-beta-galactosidase, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species F. keratolyticus
Expression System E. coli-derived msyB (Thr2-Arg124) Accession # P25738, N-terminus; GSENLYFQGHMHHHHHH; F. keratolyticus Endo-beta-Galactosidase (Asp25-Leu422) Accession # Q9ZG90, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Endo-beta-Galactosidase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.