Description
- Without calibration certificate
- For standard environments: High quality weights for regular routine testing without the need for traceability
- High-grade stainless steel: Low magnetic permeability and high corrosion resistance ensure reliable values
Specifications
Specifications
| Product Type | Test Weight |
| Material | Stainless Steel |
| Case Material | Aluminum |
| Weight (Metric) | 1 mg to 5 kg |
| Density | 8000 (± 60) kg/m3 |
| Includes | 28 Pieces; Without certificate |