Learn More
Mettler Toledo™ F2 OIML Class Advanced Weight (F2 PL C E)
Catalog No. 01912598 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Weight (Metric):
1 mg
2 mg
50 mg
1 g
20 kg
Shape:
Knob with Adjusting Cavity
Sheet
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric) Shape
01-912-598 50 mg Sheet
01-912-596 1 mg Sheet
01-912-597 2 mg Sheet
01-912-599 1 g Knob with Adjusting Cavity
01-912-600 20 kg Knob with Adjusting Cavity
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-912-598 Supplier Mettler Toledo™ Supplier No. 30406420
Add to Cart
Add to Cart
Single F2 OIML weight includes a calibration certificate and is housed in a plastic box.
  • With calibration certificate
  • Traceability Certificate: Delivered with a Calibration Certificate in accordance with ISO/IEC 17025 requirements
  • Weight Calibration Performed by Robots: State-of-the-art robot technology minimizes errors and increases accuracy
  • Weight with knob design have an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Weight
Material Stainless Steel
Case Material Plastic
Weight (Metric) 50 mg
Density 7950 (± 140) kg/m3
Shape Sheet
Includes With certificate