Learn More
Mettler Toledo™ F2 OIML Class Standard Weight (F2 PL E)
Catalog No. 01912645 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Weight (Metric):
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 kg
20 kg
Shape:
Knob with Adjusting Cavity
Sheet
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric) Shape
01-912-645 200 mg Sheet
01-912-638 1 mg Sheet
01-912-639 2 mg Sheet
01-912-640 5 mg Sheet
01-912-641 10 mg Sheet
01-912-642 20 mg Sheet
01-912-643 50 mg Sheet
01-912-644 100 mg Sheet
01-912-646 500 mg Sheet
01-912-647 1 kg Knob with Adjusting Cavity
01-912-648 20 kg Knob with Adjusting Cavity
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 01-912-645 Supplier Mettler Toledo™ Supplier No. 30402655
Add to Cart
Add to Cart
Single F2 OIML weight constructed of 304 stainless steel and housed in a plastic box.
  • Without calibration certificate
  • For Standard Environments: Affordable quality weights for regular routine testing without the need for traceability
  • Weight with knob design have an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Weight
Material Stainless Steel
Case Material Plastic
Weight (Metric) 200 mg
Density 7950 (± 140) kg/m3
Shape Sheet
Includes Without certificate