Learn More
Cell Signaling Technology F4/80 (D2S9R) XP Rabbit mAb, BSA and Azide Free, Animal-Free Manufacturing, 100 µg
SDP
Supplier:  Cell Signaling Technology 25514SF
Encompass
F4/80 (D2S9R) XP Rabbit Monoclonal Antibody is a highly specific, recombinant antibody designed for studying EMR1 in mouse models. Its BSA and Azide Free formulation allows for custom labeling with fluorophores and metals, ensuring superior lot-to-lot consistency through an animal-free manufacturing process. This product is ideal for applications such as Western Blotting and Immunohistochemistry.
Catalog No. NC2231390
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.